Learn More
Viagen Biotech Inc PROTEINASE K SOLUTION
SDP
Catalog No. NC9724952
Encompass
Proteinase K is an endolytic protease that cleaves peptide bonds at the carboxylic sides of aliphatic, aromatic or hydrophobic amino acids. Proteinase K is classified as a serine protease. The smallest peptide to be hydrolysed is a tetrapeptide. DNA Extraction lysis reagent for DNA tests & genotyping: The Proteinase K solution is useful when a small amount of biological samples are processed. Add the enzyme solution to DirectPCR reagents at the final concentration of 0.5-1.0 mg/ml (25-50 ul Proteinase K solution per 1 ml DirectPCR reagent). Follow the instruction manuals for DirectPCR Reagent Protocol/Manual for each blood, blood stain, buccal swab, hair root DNA genotyping. Once diluted in DirectPCR reagents, Proteinase K is stable for ~24 hrs.
Catalog No. NC9724952 Supplier Viagen Biotech Inc Supplier No. 501PK
Proteinase K is an endolytic protease that cleaves peptide bonds at the carboxylic sides of aliphatic, aromatic or hydrophobic amino acids. Proteinase K is classified as a serine protease. The smallest peptide to be hydrolysed is a tetrapeptide. DNA Extraction lysis reagent for DNA tests & genotyping: The Proteinase K solution is useful when a small amount of biological samples are processed. Add the enzyme solution to DirectPCR reagents at the final concentration of 0.5-1.0 mg/ml (25-50 ul Proteinase K solution per 1 ml DirectPCR reagent). Follow the instruction manuals for DirectPCR Reagent Protocol/Manual for each blood, blood stain, buccal swab, hair root DNA genotyping. Once diluted in DirectPCR reagents, Proteinase K is stable for ~24 hrs.