Learn More
Gold Biotechnology Inc Proteinase K Solution
SDP
Catalog No. 5000562030
Encompass_Preferred
Proteinase K is a highly reactive nonspecific serine protease that belongs to the subtilisin family of proteins It cleaves at the carboxylic acid side of aliphatic aromatic or hydrophobic amino acids Proteinase K is capable of inactivating RNases and DNases and is used in the isolation or preparation of high molecular weight nucleic acids Proteinase K is also useful for helping to characterize enzymes due to its cleavage specificity This enzyme was designated proteinase K because of its ability to hydrolyze keratin Proteinase K is stable in a wide variety of detergents and buffer salts and at various temperatures and pH The isoelectric point of proteinase K is 8 9
Catalog No. 50-005-62030 Supplier Gold Biotechnology Inc Supplier No. P480SL10
Proteinase K is a highly reactive nonspecific serine protease that belongs to the subtilisin family of proteins It cleaves at the carboxylic acid side of aliphatic aromatic or hydrophobic amino acids Proteinase K is capable of inactivating RNases and DNases and is used in the isolation or preparation of high molecular weight nucleic acids Proteinase K is also useful for helping to characterize enzymes due to its cleavage specificity This enzyme was designated proteinase K because of its ability to hydrolyze keratin Proteinase K is stable in a wide variety of detergents and buffer salts and at various temperatures and pH The isoelectric point of proteinase K is 8 9