Learn More
Biotang Inc PROTEINASE K SOLUTION 5 ML, P-1460, Proteinase K Solution (20mg/ml) CAS # 39450-01-6. Prepared in Tris-HCl (50 mM), calcium chloride (10 mM), and glycerol (50%). pH 8.0 +/- 0.2. Shipped cold on blue ice.
Catalog No. 50152305 Shop All BIOTANG Inc Products
Encompass_Preferred
This item has a minimum order quantity of 20 per supplier requirements. PROTEINASE K SOLUTION 5 ML, P-1460, Proteinase K Solution (20mg/ml) CAS # 39450-01-6. Prepared in Tris-HCl (50 mM), calcium chloride (10 mM), and glycerol (50%). pH 8.0 +/- 0.2. Shipped cold on blue ice.
Catalog No. 50-152-305 Supplier Biotang Inc Supplier No. BTBB852
This item has a minimum order quantity of 20 per supplier requirements. PROTEINASE K SOLUTION 5 ML, P-1460, Proteinase K Solution (20mg/ml) CAS # 39450-01-6. Prepared in Tris-HCl (50 mM), calcium chloride (10 mM), and glycerol (50%). pH 8.0 +/- 0.2. Shipped cold on blue ice.