Learn More
IBI SCIENTIFIC 3P Proteinase K Solution, 95% Purity, 5 mL
Catalog No. NC9934034
Encompass_Preferred
Proteinase K Solution - 5ml IBI Proteinase K solution is a stable 20mg/ml highly active proteolytic enzyme that is purified> br> from the mold Tritirachium album.
Catalog No. NC9934034 Supplier IBI SCIENTIFIC 3P Supplier No. IB05406
Proteinase K Solution - 5ml IBI Proteinase K solution is a stable 20mg/ml highly active proteolytic enzyme that is purified> br> from the mold Tritirachium album.