Learn More
Meridian Life Science PROTEINASE K SOLUTION 5ML
SDP
Catalog No. NC0323364
Encompass
Proteinase K is an enzyme used to digest most proteins in molecular-biological techniques. The enzyme may be used at 56C for up to 4 hours, or 37C for overnight incubations. Available as a stabilized stock solution (20mg/ml).
Catalog No. NC0323364 Supplier Meridian Life Science Supplier No. BIO37084
Proteinase K is an enzyme used to digest most proteins in molecular-biological techniques. The enzyme may be used at 56C for up to 4 hours, or 37C for overnight incubations. Available as a stabilized stock solution (20mg/ml).