Learn More
Zymo Research Corporation Proteinase K w/ Storage Buffer Set, 5mg
SDP
Catalog No. 50444112
Encompass_Preferred
The enzyme is frequently used in molecular biology applications to digest unwanted proteins such as nucleases from DNA and/or RNA preparations from microorganisms, cells, and plants.
Catalog No. 50-444-112 Supplier Zymo Research Corporation Supplier No. D300125
The enzyme is frequently used in molecular biology applications to digest unwanted proteins such as nucleases from DNA and/or RNA preparations from microorganisms, cells, and plants.