Learn More
Zymo Research Corporation Proteinase K w/ Storage Buffer Set, 60mg
SDP
Catalog No. 502037292
Encompass_Preferred
The enzyme is frequently used in molecular biology applications to digest unwanted proteins such as nucleases from DNA and/or RNA preparations from microorganisms, cells, and plants.
Catalog No. 50-203-7292 Supplier Zymo Research Corporation Supplier No. D3001260
The enzyme is frequently used in molecular biology applications to digest unwanted proteins such as nucleases from DNA and/or RNA preparations from microorganisms, cells, and plants.