Learn More
Zymo Research Corporation Proteinase K w/ Storage Buffer Set (125 mg)
SDP
Catalog No. NC1899919
Encompass_Preferred
The enzyme is frequently used in molecular biology applications to digest unwanted proteins such as nucleases from DNA and/or RNA preparations from microorganisms, cells, and plants.
Catalog No. NC1899919 Supplier Zymo Research Corporation Supplier No. D30012125
The enzyme is frequently used in molecular biology applications to digest unwanted proteins such as nucleases from DNA and/or RNA preparations from microorganisms, cells, and plants.