Learn More
ACROBiosystems Recombinant Protein G, His Tag, 2mg
Supplier:  ACROBiosystems RPGS31402MG
Encompass_Preferred
Recombinant Protein G, His Tag is fused with the polyhistidine tag at N-terminus and a single cysteine at C-terminus. The reducing (R) protein migrates as a 29 kDa polypeptide. Protein G Protein can bind to human IgG1 Fc.
Catalog No. 50-165-7789