Learn More
Proteintech Group Inc Proteintech Antibody Labeling Kit, Biotin Conjugate, Mouse IgG1, 50 Reactions
SDP
Catalog No. NC4000472 Shop All Proteintech Group Inc Products
Encompass_Preferred
This antibody labeling kit is designed for various applications, employing a FlexLinker (small polypeptide with conjugated dyes) to label unconjugated primary antibodies. Excess FlexLinker is neutralized by a FlexQuencher (Fc-containing fragment). The kit includes a PBS-based FlexBuffer and is compatible with primary antibodies in common carriers like BSA, up to 50% glycerol, and sodium azide.
  • Tested applications include immunofluorescence (IF), flow cytometry (FC), and immunohistochemistry (IHC).
  • Specific for mouse IgG1 reactivity, using biotin as the conjugate.
  • Compatible with streptavidin conjugated dyes and molecules.
  • Validated for use with antibodies in BSA, up to 50% glycerol, Tris buffer, and sodium azide.
  • Allows labeling of up to 50 different primary antibodies with a 50-reaction kit and enables scaling up.
Catalog No. NC4000472 Supplier Proteintech Group Inc Supplier No. KFA02750RXNS
Add to Cart
Add to Cart
This antibody labeling kit is designed for various applications, employing a FlexLinker (small polypeptide with conjugated dyes) to label unconjugated primary antibodies. Excess FlexLinker is neutralized by a FlexQuencher (Fc-containing fragment). The kit includes a PBS-based FlexBuffer and is compatible with primary antibodies in common carriers like BSA, up to 50% glycerol, and sodium azide.
  • Tested applications include immunofluorescence (IF), flow cytometry (FC), and immunohistochemistry (IHC).
  • Specific for mouse IgG1 reactivity, using biotin as the conjugate.
  • Compatible with streptavidin conjugated dyes and molecules.
  • Validated for use with antibodies in BSA, up to 50% glycerol, Tris buffer, and sodium azide.
  • Allows labeling of up to 50 different primary antibodies with a 50-reaction kit and enables scaling up.