Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Proteintech Group Inc PROTEINTECH GROUP INC Monoclonal Antibody, Cathepsin L, Mouse IgG2b, 150 μL
SDP

Catalog No. NC3987017 Shop All Proteintech Group Inc Products
Encompass_Preferred

This monoclonal antibody targets Cathepsin L, a lysosomal proteinase. It plays an intracellular role in normal intestinal epithelial polarization and initiation of neoplasia. Additionally, it improves cardiac function and inhibits cardiac hypertrophy, inflammation, and fibrosis by blocking AKT/GSK3B signaling.

  • Targets cathepsin L.
  • Mouse IgG2b isotype.
  • Validated for western blot (WB), immunohistochemistry (IHC), immunofluorescence/immunocytochemistry (IF/ICC), and ELISA.
  • Recognizes CTSL1 (Cathepsin L1), a lysosomal proteinase.
  • Purified using protein A purification.
  • Reactivity: human.
  • Storage at -20°C.
  • Stable for one year after shipment.
  • Detects different forms of cathepsin L (36 kDa, 39 kDa, 29 kDa and 21 kDa).
  • For research use only.
  • Available in 150 μL size.

Catalog No. NC3987017 Supplier Proteintech Group Inc Supplier No. 669141IG150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This monoclonal antibody targets Cathepsin L, a lysosomal proteinase. It plays an intracellular role in normal intestinal epithelial polarization and initiation of neoplasia. Additionally, it improves cardiac function and inhibits cardiac hypertrophy, inflammation, and fibrosis by blocking AKT/GSK3B signaling.

  • Targets cathepsin L.
  • Mouse IgG2b isotype.
  • Validated for western blot (WB), immunohistochemistry (IHC), immunofluorescence/immunocytochemistry (IF/ICC), and ELISA.
  • Recognizes CTSL1 (Cathepsin L1), a lysosomal proteinase.
  • Purified using protein A purification.
  • Reactivity: human.
  • Storage at -20°C.
  • Stable for one year after shipment.
  • Detects different forms of cathepsin L (36 kDa, 39 kDa, 29 kDa and 21 kDa).
  • For research use only.
  • Available in 150 μL size.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.