Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Proteintech Group Inc Proteintech Group Polyclonal Antibody, Rabbit/IgG, Human, WB/IHC/IP/ELISA
SDP

Catalog No. NC3990068 Shop All Proteintech Group Inc Products
Encompass

The TRMT112 Polyclonal antibody targets TRMT112 and is validated for use in WB, IHC, IP, and ELISA applications, showing reactivity with human samples. It is a small, evolutionarily conserved protein that acts as a co-factor and activator for various MTases involved in rRNA, tRNA, and protein methylation. The interaction of TRMT112 with 18S rRNA MTases WBSCR22 (BUD23, MERM1) and METTL5 is essential for their metabolic stability and enzymatic activity in cells.

  • Validated for western blot (WB), immunohistochemistry (IHC), immunoprecipitation (IP), and ELISA applications.
  • Shows reactivity with human samples.
  • Host/isotype: Rabbit / IgG.
  • Conjugation type: Unconjugated.
  • Purification method: Antigen affinity purification.
  • Buffer formulation: PBS with 0.02% sodium azide and 50% glycerol, pH 7.3.
  • Storage conditions: Store at -20°C. Stable for one year after shipment.

Catalog No. NC3990068 Supplier Proteintech Group Inc Supplier No. 264721AP
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The TRMT112 Polyclonal antibody targets TRMT112 and is validated for use in WB, IHC, IP, and ELISA applications, showing reactivity with human samples. It is a small, evolutionarily conserved protein that acts as a co-factor and activator for various MTases involved in rRNA, tRNA, and protein methylation. The interaction of TRMT112 with 18S rRNA MTases WBSCR22 (BUD23, MERM1) and METTL5 is essential for their metabolic stability and enzymatic activity in cells.

  • Validated for western blot (WB), immunohistochemistry (IHC), immunoprecipitation (IP), and ELISA applications.
  • Shows reactivity with human samples.
  • Host/isotype: Rabbit / IgG.
  • Conjugation type: Unconjugated.
  • Purification method: Antigen affinity purification.
  • Buffer formulation: PBS with 0.02% sodium azide and 50% glycerol, pH 7.3.
  • Storage conditions: Store at -20°C. Stable for one year after shipment.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.