Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Proteintech Group Inc Proteintech Monoclonal Antibody, Cleaved PARP1, 1000 μg/mL, 150 μL, Clone 4G4C8
SDP

Catalog No. NC3990534 Shop All Proteintech Group Inc Products
Encompass

This monoclonal antibody specifically recognizes the cleaved form of PARP1, a nuclear enzyme involved in DNA damage repair. Cleaved PARP1 fragments are common biomarkers of apoptosis, resulting from caspase activation during the apoptotic process. This antibody does not recognize full-length PARP1.

  • Specifically recognizes cleaved PARP1
  • Clone 4G4C8
  • Host species: Mouse
  • Isotype: IgG1
  • Reactivity: Human, mouse, rat
  • Applications: Western Blot, Immunohistochemistry, Immunofluorescence/ICC, Flow Cytometry (Intra), ELISA
  • Concentration: 1000 μg/mL
  • Volume: 150 μL
  • Formulation: PBS with 0.02% sodium azide and 50% glycerol, pH 7.3
  • Unconjugated
  • Purification method: Protein G purification
  • Store at -20°C; stable for one year after shipment

Catalog No. NC3990534 Supplier Proteintech Group Inc Supplier No. 605551IG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This monoclonal antibody specifically recognizes the cleaved form of PARP1, a nuclear enzyme involved in DNA damage repair. Cleaved PARP1 fragments are common biomarkers of apoptosis, resulting from caspase activation during the apoptotic process. This antibody does not recognize full-length PARP1.

  • Specifically recognizes cleaved PARP1
  • Clone 4G4C8
  • Host species: Mouse
  • Isotype: IgG1
  • Reactivity: Human, mouse, rat
  • Applications: Western Blot, Immunohistochemistry, Immunofluorescence/ICC, Flow Cytometry (Intra), ELISA
  • Concentration: 1000 μg/mL
  • Volume: 150 μL
  • Formulation: PBS with 0.02% sodium azide and 50% glycerol, pH 7.3
  • Unconjugated
  • Purification method: Protein G purification
  • Store at -20°C; stable for one year after shipment

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.