Learn More
Proteintech Group Inc Proteintech Recombinant Monoclonal Antibody, RAB7A, Rabbit IgG, Unconjugated
SDP
Catalog No. NC3991866 Shop All Proteintech Group Inc Products
Encompass_Preferred
This recombinant monoclonal antibody targets RAB7A, a small GTPase superfamily and Rab family member. It is involved in late endocytic transport and contributes to the maturation of phagosomes. Defects in RAB7A are associated with Charcot–Marie–Tooth disease type 2B (CMT2B).
  • Specific to RAB7A.
  • Provided as a rabbit IgG.
  • Purified via protein A purification.
  • Unconjugated form.
  • Stable for one year when stored at –20°C.
  • Suitable for Western blot, immunohistochemistry, immunofluorescence/ICC, flow cytometry (intracellular), and ELISA.
  • Tested for reactivity in human, mouse, and rat samples.
Catalog No. NC3991866 Supplier Proteintech Group Inc Supplier No. 847411RR100UL
This recombinant monoclonal antibody targets RAB7A, a small GTPase superfamily and Rab family member. It is involved in late endocytic transport and contributes to the maturation of phagosomes. Defects in RAB7A are associated with Charcot–Marie–Tooth disease type 2B (CMT2B).
  • Specific to RAB7A.
  • Provided as a rabbit IgG.
  • Purified via protein A purification.
  • Unconjugated form.
  • Stable for one year when stored at –20°C.
  • Suitable for Western blot, immunohistochemistry, immunofluorescence/ICC, flow cytometry (intracellular), and ELISA.
  • Tested for reactivity in human, mouse, and rat samples.