Learn More
Waters Corp Sample Preparation Kit, High Digest, Ambient Refill, 96 Analysis
Catalog No. 502391299 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This sample preparation workflow streamlines complex pre-analytic procedures for LC-MS protein quantification using the surrogate peptide technique. It is designed for high protein content samples (1.0 to 5.25 mg total protein per sample), directly degrading proteins in whole, non-immuno-purified plasma or serum. The kit aims to shorten method development time and provide repeatable, accurate, and sensitive protein measurement.
  • Includes pre-measured, lot-traceable digestion buffer, trypsin dissolving reagent, digestion inactivation reagent, and a RapiGest denaturant vial.
  • Designed for high protein content samples (1.0 to 5.25 mg total protein per sample).
  • Supports both 3-step or 5-step protocols for protein quantification.
  • Optimized for LC-MS/MS detection and RPLC separation.
Catalog No. 50-239-1299 Supplier Waters Corp Supplier No. 186008872
This sample preparation workflow streamlines complex pre-analytic procedures for LC-MS protein quantification using the surrogate peptide technique. It is designed for high protein content samples (1.0 to 5.25 mg total protein per sample), directly degrading proteins in whole, non-immuno-purified plasma or serum. The kit aims to shorten method development time and provide repeatable, accurate, and sensitive protein measurement.
  • Includes pre-measured, lot-traceable digestion buffer, trypsin dissolving reagent, digestion inactivation reagent, and a RapiGest denaturant vial.
  • Designed for high protein content samples (1.0 to 5.25 mg total protein per sample).
  • Supports both 3-step or 5-step protocols for protein quantification.
  • Optimized for LC-MS/MS detection and RPLC separation.