Learn More
Waters Corp WATERS CORPORATION ProteinWorks Auto-eXpress High 5 Digest Kit, 96 Analysis
Supplier: Waters Corp 176004080
The ProteinWorks™ Auto-eXpress High 5 Digest Kit simplifies and accelerates digestion of plasma & serum samples for LC-MS protein quantification in bioanalysis. It offers an automation-friendly protocol with pre-measured, lot-traceable reagents. Suitable for high protein content samples (1.0 to 5.25 mg total protein), it is compatible with purified proteins, cell lysates, and tissue extracts, optimized for LC-MS systems.
- 96 analyses per kit.
- RPLC separation and MS/MS detection for protein/peptide analysis.
- Automated liquid handling optimization reduces hands-on time.
- Compatible with purified proteins, cell lysates, tissue extracts.
- For samples with 1.0 to 5.25 mg total protein.
- 5-step protocol: denaturation, reduction, alkylation, digestion, quench.
- Supports standard (205 μL) and low volume (164 μL) protocols.