Learn More
Waters Corp Waters Digest Kit, ProteinWorks Auto-eXpress High 3, Plasma/Serum, 96 Analyses
Catalog No. 501528566 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit simplifies and accelerates digestion of whole plasma and serum samples for LC-MS protein quantification. It includes a robust protocol and pre-measured, lot-traceable reagents in an automation friendly format. Optimized for denaturation, digestion, and quench of proteins in biological matrices, it handles samples with 1.0 to 5.25 mg total protein.
  • Simplifies and accelerates digestion of plasma/serum samples.
  • Enables LC-MS protein quantification via surrogate peptide approach.
  • Includes robust digest protocol and pre-measured, lot-traceable reagents.
  • Automation friendly format.
  • Supports denaturation, digestion, and quench of proteins.
  • Optimized for 1.0 to 5.25 mg total protein.
  • Accommodates 32 to 96 samples per analysis.
  • Utilizes a 3-step protocol: denaturation, digestion, quench.
Catalog No. 50-152-8566 Supplier Waters Corp Supplier No. 176004079
This kit simplifies and accelerates digestion of whole plasma and serum samples for LC-MS protein quantification. It includes a robust protocol and pre-measured, lot-traceable reagents in an automation friendly format. Optimized for denaturation, digestion, and quench of proteins in biological matrices, it handles samples with 1.0 to 5.25 mg total protein.
  • Simplifies and accelerates digestion of plasma/serum samples.
  • Enables LC-MS protein quantification via surrogate peptide approach.
  • Includes robust digest protocol and pre-measured, lot-traceable reagents.
  • Automation friendly format.
  • Supports denaturation, digestion, and quench of proteins.
  • Optimized for 1.0 to 5.25 mg total protein.
  • Accommodates 32 to 96 samples per analysis.
  • Utilizes a 3-step protocol: denaturation, digestion, quench.