Learn More
Waters Corp ProteinWorks Digest Kit, 96 Analysis, LC-MS System, Low Protein Content
Catalog No. 501528565 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This ProteinWorks™ Auto-eXpress Low 5 Digest Kit simplifies and accelerates LC-MS protein quantification in immunopurified plasma & serum samples. It provides pre-measured, lot-traceable reagents and a robust digest protocol in an automation-friendly format. Suitable for low protein content samples (0.2 – 1.0 mg total protein).
  • Simplifies and accelerates LC-MS protein quantification.
  • Robust digest protocol and pre-measured reagents.
  • Automation-friendly format.
  • Suitable for low protein content (0.2 – 1.0 mg total protein).
  • Ensures precise, accurate, and reliable quantification.
Catalog No. 50-152-8565 Supplier Waters Corp Supplier No. 176004078
This ProteinWorks™ Auto-eXpress Low 5 Digest Kit simplifies and accelerates LC-MS protein quantification in immunopurified plasma & serum samples. It provides pre-measured, lot-traceable reagents and a robust digest protocol in an automation-friendly format. Suitable for low protein content samples (0.2 – 1.0 mg total protein).
  • Simplifies and accelerates LC-MS protein quantification.
  • Robust digest protocol and pre-measured reagents.
  • Automation-friendly format.
  • Suitable for low protein content (0.2 – 1.0 mg total protein).
  • Ensures precise, accurate, and reliable quantification.