Learn More
Waters Corp ProteinWorks eXpress Digest Kit, LC-MS Protein Quantification, 96 Analysis
Catalog No. 502392034 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ProteinWorks eXpress Digest Start-up Kit is designed to simplify and accelerate the direct digestion of immunopurified plasma & serum samples for LC-MS protein quantification using the surrogate peptide approach.
  • Simplifies and accelerates direct digestion for LC-MS protein quantification of immunopurified plasma & serum samples.
  • Provides a standardized, proven route for fast, accurate, and precise protein quantification in bioanalysis.
  • Includes pre-measured, lot-traceable chemicals optimized for dependable LC-MS protein quantification.
  • Enables rapid quantification, such as Infliximab down to 10 ng/mL.
  • Achieves single-digit intra-kit and inter-kit reproducibility across multiple monoclonal antibodies.
Catalog No. 50-239-2034 Supplier Waters Corp Supplier No. 176003689
The ProteinWorks eXpress Digest Start-up Kit is designed to simplify and accelerate the direct digestion of immunopurified plasma & serum samples for LC-MS protein quantification using the surrogate peptide approach.
  • Simplifies and accelerates direct digestion for LC-MS protein quantification of immunopurified plasma & serum samples.
  • Provides a standardized, proven route for fast, accurate, and precise protein quantification in bioanalysis.
  • Includes pre-measured, lot-traceable chemicals optimized for dependable LC-MS protein quantification.
  • Enables rapid quantification, such as Infliximab down to 10 ng/mL.
  • Achieves single-digit intra-kit and inter-kit reproducibility across multiple monoclonal antibodies.