Learn More
Waters Corp ProteinWorks eXpress Digest Start-up Kit, 96 Analyses, Protein Quantification, LC-MS
Catalog No. 502392150 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit simplifies and accelerates direct digestion of immunopurified plasma and serum samples for LC-MS protein quantification. It includes pre-measured, lot-traceable reagents, a 96-tube sample collection module, and optimized protocols for accurate protein quantification.
  • Accelerates direct digestion of immunopurified plasma and serum samples for LC-MS protein quantification.
  • Contains pre-measured, lot-traceable reagents and optimized protocols.
  • Offers a 96-tube sample collection module for 96 analyses.
  • Delivers single-digit intra-kit and inter-kit reproducibility for monoclonal antibodies.
  • Increases assay sensitivity and system resilience by removing excess digest reagents and plasma/serum components.
  • Ensures laboratory and data consistency via a validated, standardized technique.
Catalog No. 50-239-2150 Supplier Waters Corp Supplier No. 176003696
This kit simplifies and accelerates direct digestion of immunopurified plasma and serum samples for LC-MS protein quantification. It includes pre-measured, lot-traceable reagents, a 96-tube sample collection module, and optimized protocols for accurate protein quantification.
  • Accelerates direct digestion of immunopurified plasma and serum samples for LC-MS protein quantification.
  • Contains pre-measured, lot-traceable reagents and optimized protocols.
  • Offers a 96-tube sample collection module for 96 analyses.
  • Delivers single-digit intra-kit and inter-kit reproducibility for monoclonal antibodies.
  • Increases assay sensitivity and system resilience by removing excess digest reagents and plasma/serum components.
  • Ensures laboratory and data consistency via a validated, standardized technique.