Learn More
Waters Corp ProteinWorks eXpress Direct Digest Start-up Kit, Plasma & Serum Samples, LC-MS, 96 Analyses
Catalog No. 502391901 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit simplifies and accelerates the direct digestion of whole, non-immunopurified plasma & serum samples for LC-MS protein quantification. It offers a standardized, proven method for fast, accurate, and precise protein quantification in bioanalysis.
  • Simplified and accelerated digestion of plasma and serum samples.
  • Comprehensive kit components for complete workflow.
  • Standardized and reproducible results with lot-traceable reagents.
  • Flexible format, analyzing 8 to 96 samples per run.
  • Optimized for LC-MS protein quantification.
  • Accommodates diverse proteins with 3-step and 5-step protocols.
  • Includes essential digestion, reduction, alkylation, and inactivation agents.
Catalog No. 50-239-1901 Supplier Waters Corp Supplier No. 176003695
This kit simplifies and accelerates the direct digestion of whole, non-immunopurified plasma & serum samples for LC-MS protein quantification. It offers a standardized, proven method for fast, accurate, and precise protein quantification in bioanalysis.
  • Simplified and accelerated digestion of plasma and serum samples.
  • Comprehensive kit components for complete workflow.
  • Standardized and reproducible results with lot-traceable reagents.
  • Flexible format, analyzing 8 to 96 samples per run.
  • Optimized for LC-MS protein quantification.
  • Accommodates diverse proteins with 3-step and 5-step protocols.
  • Includes essential digestion, reduction, alkylation, and inactivation agents.