Learn More
Enzo Life Sciences PROTEOSTAT Protein refolding and aggregation sensing kit (2x96wells)
SDP
Catalog No. NC1032830
Encompass_Preferred
Identify Optimal Protein Refolding Conditions and Maximize Recovery of Functional Protein. Contents: PROTEOSTAT Detection Reagent, 22 µl. 10X PROTEOSTAT Assay Buffer, 1 x 2 ml. 2X Refolding Buffers 1-15, 10 ml each. Lysozyme standard, 10 mg. Denaturant: 6M Guanidine Hydrochloride (GdnHCl), Tris-HCl (pH 8), 14 ml. 100 mM DTT, 1 ml. GSH, 46.1 mg. GSSG, 18.4 mg. 10 mM PEG, 1 ml. 500 mM EDTA, 1 ml. 400 mM CaCl2, 1 ml. 400 mM MgCl2, 1 ml. 1 M NaCl, 1 ml. 0.1% Tween-20, 1 ml. 1.2 M Sucrose, 1 ml. α-Cyclodextrin  (200 mg/ml), 1 ml
Catalog No. NC1032830 Supplier Enzo Life Sciences Supplier No. ENZ51040KP002
Identify Optimal Protein Refolding Conditions and Maximize Recovery of Functional Protein. Contents: PROTEOSTAT Detection Reagent, 22 µl. 10X PROTEOSTAT Assay Buffer, 1 x 2 ml. 2X Refolding Buffers 1-15, 10 ml each. Lysozyme standard, 10 mg. Denaturant: 6M Guanidine Hydrochloride (GdnHCl), Tris-HCl (pH 8), 14 ml. 100 mM DTT, 1 ml. GSH, 46.1 mg. GSSG, 18.4 mg. 10 mM PEG, 1 ml. 500 mM EDTA, 1 ml. 400 mM CaCl2, 1 ml. 400 mM MgCl2, 1 ml. 1 M NaCl, 1 ml. 0.1% Tween-20, 1 ml. 1.2 M Sucrose, 1 ml. α-Cyclodextrin  (200 mg/ml), 1 ml