Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ ProteoChrom™ HPTLC Cellulose Plate

Catalog No. M1056510001 Shop All MilliporeSigma Products
Catalog No. M1056510001 Supplier MilliporeSigma™ Supplier No. 1.05651.0001

missing translation for 'validMainframeMsg'

Used for high performance thin layer chromatography

  • Separates proteins
  • Used with aqueous solvent systems
  • Stable in water
  • Layer thickness: 90 to 120μm
  • Separation number: >11
  • Size: 10 x 10cm
TRUSTED_SUSTAINABILITY

Specifications

Color White
Material Cellulose
Type Cellulose Plate
For Use With (Application) Thin Layer Chromatography
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.