Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ ProteoExtract™ Subcellular Proteome Extraction Kit

Catalog No. 5397901KIT Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-979-01KIT 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-979-01KIT Supplier MilliporeSigma™ Supplier No. 5397901KIT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed for fast and reproducible extraction of subcellular proteomes from adherent and suspension-grown mammalian cells

  • Stepwise extraction results in four distinct subcellular proteomes from one sample
  • Highly reproducible
  • No ultracentrifugation steps
  • Fast—needs just 2 hours with 45 minutes hands-on time
  • Produces proteins suitable for functional studies
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 kit
Format 20 Extractions
Kit Contents Wash buffer, Extraction buffer I, II, III, IV, Protease inhibitor cocktail, Benzonase nuclease, and a user protocol
Content And Storage Store at+2°C to+8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.