Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProteoMass™ ACTH Fragment 18-39 MALDI-MS Standard, MilliporeSigma™ Supelco™

Catalog No. 111014531 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 x 1 Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4531 5 x 1 Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4531 Supplier MilliporeSigma Supelco Supplier No. A83465X1VL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The single peptide or protein standard is used to calibrate and test matrix assisted laser desorption ionization (MALDI) mass spectrometers. Adrenocorticotropic hormone (ACTH) is synthesized from its precursor pro-opiomelanocortin (POMC). It is released from anterior pituitary gland in response to corticotropin-releasing hormone (CRH) from hypothalamus.
TRUSTED_SUSTAINABILITY

Specifications

Packaging Vial
CAS 53917-42-3
Product Line ProteoMass
MDL Number MFCD00076159
Synonym Adrenocorticotropic hormone fragment 18-39
Type MALDI-MS Standard
Quantity 5 x 1 Vial

ProteoMass is a trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.