Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProteoMass™ Albumin MALDI-MS Standard, MilliporeSigma™ Supelco™

Catalog No. 111004055 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 x 10 nmol Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-4055 5 x 10 nmol Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-4055 Supplier MilliporeSigma Supelco Supplier No. A84715X1VL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Packaging Vial
Product Line ProteoMass
MDL Number MFCD00130384
Synonym Albumin, bovine, mass spec standard
Type MALDI-MS Standard
Quantity 5 x 10 nmol Vial

ProteoMass is a trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.