Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Proteome Profiler Mouse XL Cytokine Array
GSA_VA

Catalog No. ary028
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ARY028 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. ARY028 Supplier R&D Systems™ Supplier No. ARY028
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

The Proteome Profiler Mouse XL Cytokine Array requires no specialized equipment and eliminates the need for multiple Western blot experiments.

The Proteome Profiler Mouse XL Cytokine Array contains 4 membranes-each spotted in duplicate with 111 different cytokine antibodies.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

For Use With (Application) For parallel determination of mouse cytokines and chemokines
Product Type Cytokine Array Kits
Quantity 25 μg
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.