Learn More
R&D Systems™ Proteome Purify 2 Human Serum Protein Immunodepletion Resin
Catalog No. IDR002020
Change view
Click to view available options
Quantity:
20 Tests
40 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IDR002020 20 Tests
IDR002040 40 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. IDR002020 Supplier R&D Systems™ Supplier No. IDR002020
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
Immunodepletion Resin, Spin-X Filter Units
A single-use resin that depletes serum or plasma of albumin and IgG to improve visualization of target proteins
Allows user to increase protein load or concentration
Removal allows visualization of proteins that co-migrate or co-purify with the high-abundance proteins
Kit removes proteins representing approximately 60% of the protein contained in serum and plasma
Specifications
Quantity 20 Tests
Product Type Immunodepletion Resin
Content And Storage Store the unopened product at 2 - 8° C. Do not use past expiration date.