Learn More
AB Sciex LLC PROTEOMELAB IgG Control Standard 3 Pack
Catalog No. NC9116585
Encompass
IgG Control Standard 3 Pack. The IgG Control Standard allows for experimental control when separating reduced or non-reduced IgG. For research use only. Not for use in diagnostic procedures.
Catalog No. NC9116585 Supplier AB Sciex LLC Supplier No. 391734
IgG Control Standard 3 Pack. The IgG Control Standard allows for experimental control when separating reduced or non-reduced IgG. For research use only. Not for use in diagnostic procedures.