Learn More
SMART TUBE INC Proteomic Stabilizer PROT1, size: 1 Liter
Catalog No. 501351691
Encompass_Preferred
1 liter bottle of Proteomic Stabilizer PROT1. Sufficient to stabilize over 700 milliliters of whole blood when used as recommended. Also includes buffer used after thawing samples stored frozen in Stabilizer: THAWLYSE1 Thaw-Lyse, 1000X Concentrate. User information must be added to all orders for accurate processing
This item is not returnable. View return policy
Catalog No. 501351691 Supplier SMART TUBE INC Supplier No. PROT11L
1 liter bottle of Proteomic Stabilizer PROT1. Sufficient to stabilize over 700 milliliters of whole blood when used as recommended. Also includes buffer used after thawing samples stored frozen in Stabilizer: THAWLYSE1 Thaw-Lyse, 1000X Concentrate. User information must be added to all orders for accurate processing