Learn More
SMART TUBE INC Proteomic Stabilizer PROT1, size: 100 Tests
Catalog No. 501351689
Encompass
Bottle of Proteomic Stabilizer. Volume sufficient for stabilizing one hundred 0.5ml samples of whole blood. Also includes buffer used after thawing samples stored frozen in Stabilizer: THAWLYSE1 Thaw-Lyse, 1000X Concentrate. User information must be added to all orders for accurate processing
This item is not returnable. View return policy
Catalog No. 501351689 Supplier SMART TUBE INC Supplier No. PROT1
Bottle of Proteomic Stabilizer. Volume sufficient for stabilizing one hundred 0.5ml samples of whole blood. Also includes buffer used after thawing samples stored frozen in Stabilizer: THAWLYSE1 Thaw-Lyse, 1000X Concentrate. User information must be added to all orders for accurate processing