Learn More
Sigma Aldrich Fine Chemicals Biosciences Proteomics Dynamic Range S
Supplier:  Sigma Aldrich Fine Chemicals Biosciences UPS21SET
Encompass
The Proteomics Dynamic Range Standard Set is produced from a mixture of 48 individual human source or human sequence recombinant proteins each of which has been selected to limit heterogeneous post-translational modifications (PTMs). The protein standard is formulated from 6 mixtures of 8 proteins to present a dynamic range of 5 orders of magnitude ranging from 50 pmoles to 500 amoles. Each protein has been quantitated by amino acid analysis (AAA) prior to formulation.
Catalog No. 50-291-4689