Learn More
Research Products International Corp Proteose Peptone 500 G
SDP
Catalog No. 5000052403
Encompass_Preferred
Animal-derived peptone is a protein hydrolysate that supplies peptides, amino acids, and nitrogen for microbial growth. Commonly used as a nutrient supplement in culture media, it supports the cultivation of diverse bacterial and fungal species. Its consistent composition enables reliable results in clinical, industrial, and research microbiology.
Catalog No. 50-000-52403 Supplier Research Products International Corp Supplier No. P32750500.0
Animal-derived peptone is a protein hydrolysate that supplies peptides, amino acids, and nitrogen for microbial growth. Commonly used as a nutrient supplement in culture media, it supports the cultivation of diverse bacterial and fungal species. Its consistent composition enables reliable results in clinical, industrial, and research microbiology.