Learn More
Sigma Aldrich Fine Chemicals Biosciences ProteoSilverTM Silver Stai
Catalog No. 502904136
Encompass
Conveniently packaged extremely stable and highly sensitive ProteoSilver is an ideal product for any protein scientist. The kit contains prepared SOLUTIONons of silver staining reagents along with detailed instructions to achieve optimal results. With a detection limit of 0.1 ng/mm2 of protein (BSA) and an extremely low background ProteoSilver leads to superior detection of low abundance proteins.
Catalog No. 50-290-4136 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. PROTSIL11KT
Conveniently packaged extremely stable and highly sensitive ProteoSilver is an ideal product for any protein scientist. The kit contains prepared SOLUTIONons of silver staining reagents along with detailed instructions to achieve optimal results. With a detection limit of 0.1 ng/mm2 of protein (BSA) and an extremely low background ProteoSilver leads to superior detection of low abundance proteins.