Learn More
Bioworld PROTEOSPIN DETERGENT CLEAN-
Catalog No. 50198473
Encompass_Preferred
PROTEOSPIN DETERGENT CLEAN-UP MAXI KIT: For fast and simple removal of detergents from protein samples Concentrate protein samples and remove detergents at the same time; Rapid and convenient spin-column protocol; Effectively remove a w...
Catalog No. 50-198-473 Supplier Bioworld Supplier No. 208400291
PROTEOSPIN DETERGENT CLEAN-UP MAXI KIT: For fast and simple removal of detergents from protein samples Concentrate protein samples and remove detergents at the same time; Rapid and convenient spin-column protocol; Effectively remove a w...