Learn More
Bioworld PROTEOSPIN INCLUSION BODY I
Catalog No. 50198476
Encompass_Preferred
PROTEOSPIN INCLUSION BODY ISOLATION MAXI KIT: For the rapid isolation of inclusion body proteins All-in-one solution for inclusion body protein isolation and purifcation; Complete kitwith Cell Lysis Reagent, Inclusion Body Solubilizatio...
Catalog No. 50-198-476 Supplier Bioworld Supplier No. 208400321
PROTEOSPIN INCLUSION BODY ISOLATION MAXI KIT: For the rapid isolation of inclusion body proteins All-in-one solution for inclusion body protein isolation and purifcation; Complete kitwith Cell Lysis Reagent, Inclusion Body Solubilizatio...