Learn More
Bioworld ProteoSpin On-Column Proteolytic Digestion Kit, 25 Prep(s)
Catalog No. 50198479
Encompass_Preferred
For the rapid and convenient on-column digestion of proteins Fast and simple procedure to generate digested peptides Simultaneous digestion and concentration Peptides are ready for applications such as mass spectrometry and SDS-PAGEFEATURES AND BENEFITSFast method for generating peptides - Compared to traditional methods of peptide generation and purification, the ProteoSpin™ On-Column Proteolytic Digestion kit is very rapid. Does not generate artifacts - Peptide generation is complete, with no generation of additional artifacts being detected in mass spectrometry. Digestion and purification in a single column - The use of multiple columns and tubes for digestion and purification is avoided; lower cost and increased convenience for user.
Catalog No. 50-198-479 Supplier Bioworld Supplier No. 208400351
For the rapid and convenient on-column digestion of proteins Fast and simple procedure to generate digested peptides Simultaneous digestion and concentration Peptides are ready for applications such as mass spectrometry and SDS-PAGEFEATURES AND BENEFITSFast method for generating peptides - Compared to traditional methods of peptide generation and purification, the ProteoSpin™ On-Column Proteolytic Digestion kit is very rapid. Does not generate artifacts - Peptide generation is complete, with no generation of additional artifacts being detected in mass spectrometry. Digestion and purification in a single column - The use of multiple columns and tubes for digestion and purification is avoided; lower cost and increased convenience for user.