Learn More
Bioworld PROTEOSPIN TOTAL PROTEIN DE
Supplier:  Bioworld 208400371
Encompass_Preferred
PROTEOSPIN TOTAL PROTEIN DETERGENT CLEANUP KIT: For the rapid concentration and clean-up of protein samples Concentrate protein samples and remove detergents at the same time; Rapid and convenient spin-column protocol; Effectively remov...
Catalog No. 50-198-481