Learn More
Bioworld PROTEOSPIN URINE PROTEIN CO
Catalog No. 50198477
Encompass_Preferred
PROTEOSPIN URINE PROTEIN CONCENTRATION KIT: For the rapid and efficient concentration of total proteins from urine samples Simultaneous concentration, desalting and buffer exchange of total urinary proteins; No molecular weight cutoff, ...
Catalog No. 50-198-477 Supplier Bioworld Supplier No. 208400331
PROTEOSPIN URINE PROTEIN CONCENTRATION KIT: For the rapid and efficient concentration of total proteins from urine samples Simultaneous concentration, desalting and buffer exchange of total urinary proteins; No molecular weight cutoff, ...