Learn More
Enzo Life Sciences PROTEOSTAT PDI assay kit (50tests)
SDP
Catalog No. 502012594
Encompass_Preferred
The PROTEOSTAT PDI assay kit provides a simple, homogenous assay for screening modulators of PDI enzymatic activity. This is accomplished by monitoring PDI-catalyzed reduction of insulin in the presence of Dithiothreitol (DTT), resulting in the formation of insulin aggregates which then bind avidly to the red-emitting fluorogenic PROTEOSTAT PDI detection dye. Relative to analogous turbidimetric assays of PDI activity, the fluorescence-based assay provides a vastly improved assay signal window, improved lower detection limit, and superior Z’-score (>0.8). Intra-plate and inter-plate CVs using the assay are typically 3-5%. Human recombinant PDI enzyme (EC 5.3.4.1) and the PDI inhibitor, bacitracin, are included in the kit, as well as all reagents necessary for monitoring changes in PDI activity. Alternative name: Protein disulfide isomerase. Applications: Fluorescent detection .
Catalog No. 50-201-2594 Supplier Enzo Life Sciences Supplier No. ENZ51024KP050
The PROTEOSTAT PDI assay kit provides a simple, homogenous assay for screening modulators of PDI enzymatic activity. This is accomplished by monitoring PDI-catalyzed reduction of insulin in the presence of Dithiothreitol (DTT), resulting in the formation of insulin aggregates which then bind avidly to the red-emitting fluorogenic PROTEOSTAT PDI detection dye. Relative to analogous turbidimetric assays of PDI activity, the fluorescence-based assay provides a vastly improved assay signal window, improved lower detection limit, and superior Z’-score (>0.8). Intra-plate and inter-plate CVs using the assay are typically 3-5%. Human recombinant PDI enzyme (EC 5.3.4.1) and the PDI inhibitor, bacitracin, are included in the kit, as well as all reagents necessary for monitoring changes in PDI activity. Alternative name: Protein disulfide isomerase. Applications: Fluorescent detection .