Learn More
Enzo Life Sciences PROTEOSTAT Thioredoxin-1 assay kit (2x96wells)
SDP
Supplier:  Enzo Life Sciences ENZ51033KP002
Encompass_Preferred
Applications: Fluorescent detection. Contents: PROTEOSTAT PDI detection reagent: 20 μL. Thioredoxin-1 (human) (recombinant): 2 vials x 80 μL. Insulin: 2 vials x 1.8 μmol. Aurothiomalate (inhibitor): 2 vials x 25 nmol. PBE buffer: 25 mL. Stop reagent: 1 mL. DTT: 2 vials x 1.3 mL. Deionized water: 5 mL
Catalog No. NC1577355