Learn More
Enzo Life Sciences PROTEOSTAT Protein aggregation assay (2x96tests)
SDP
Catalog No. NC0005516
Encompass_Preferred
Quantitative Detection of  Protein Aggregates from Visible to Subvisible Particles. PROTEOSTAT.0.5). Lyophilized native and aggregated protein are provided as negative and positive controls for monitoring changes in protein aggregation status.PROTEOSTAT.Easy to use - simply add water!. Applications: Flow Cytometry, Fluorescence microscopy, Fluorescent detection, Microplate. Contents: PROTEOSTAT detection reagent. PROTEOSTAT positive control. PROTEOSTAT negative control. 10X PROTEOSTAT assay buffer
Catalog No. NC0005516 Supplier Enzo Life Sciences Supplier No. ENZ51023KP002
Quantitative Detection of  Protein Aggregates from Visible to Subvisible Particles. PROTEOSTAT.0.5). Lyophilized native and aggregated protein are provided as negative and positive controls for monitoring changes in protein aggregation status.PROTEOSTAT.Easy to use - simply add water!. Applications: Flow Cytometry, Fluorescence microscopy, Fluorescent detection, Microplate. Contents: PROTEOSTAT detection reagent. PROTEOSTAT positive control. PROTEOSTAT negative control. 10X PROTEOSTAT assay buffer