Learn More
ATCC Proteus hauseri
Catalog No. 502383768
Encompass
Proteus hauseri NCTC 4175 strain Lehmann is a whole-genome sequenced bacterial strain with applications in quality control and water testing.
Catalog No. 50-238-3768 Supplier ATCC Supplier No. 13315
Proteus hauseri NCTC 4175 strain Lehmann is a whole-genome sequenced bacterial strain with applications in quality control and water testing.