Learn More
ATCC Proteus mirabilis
Supplier:  ATCC 29906
Encompass
Proteus mirabilis strain CDC PR 14 is a whole-genome sequenced type strain. This bacterial culture can be used for media testing and it is recommended by ATCC for use in the tests described in ASTM Standard Test Method E979-91 where only the taxon is specified.
Catalog No. 50-238-2377