Learn More
ATCC Proteus mirabilis
Supplier:  ATCC 25933
Encompass
Proteus mirabilis Hauser strain NCDC 2059-70 was isolated from a human vagina. This whole-genome sequenced bacterium can be used as a quality control strain for IDS products.
Catalog No. 50-238-3806
 Restricted Item