Learn More
Viva Products NoEndoTM Endotoxin Removal Spin Columns, High Capacity
SDP
Catalog No. NC2086461
Encompass
Proteus NoEndoTM Spin Column Kit; For fast affinity based removal of endotoxins from antibodies and other protein samples; 20 ml capacity; High Capacity resin can remove over 500,000 EU of endotoxin; 12/PK
This item is not returnable. View return policy
Catalog No. NC2086461 Supplier Viva Products Supplier No. GENNOE12HC
Proteus NoEndoTM Spin Column Kit; For fast affinity based removal of endotoxins from antibodies and other protein samples; 20 ml capacity; High Capacity resin can remove over 500,000 EU of endotoxin; 12/PK