Learn More
Viva Products NoEndoTM Endotoxin Removal Spin Columns, Standard Capacity
SDP
Catalog No. NC0688150
Encompass
Proteus NoEndoTM Spin Column Kit; For fast affinity based removal of endotoxins from antibodies and other protein samples; 20 ml capacity; Standard Capacity resin can remove 30,000 EU of endotoxin; 48/PK
Catalog No. NC0688150 Supplier Viva Products Supplier No. GENNOE48S
Proteus NoEndoTM Spin Column Kit; For fast affinity based removal of endotoxins from antibodies and other protein samples; 20 ml capacity; Standard Capacity resin can remove 30,000 EU of endotoxin; 48/PK