Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Proteus penneri Z527, titered
SDP

Catalog No. 22158383 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-158-383 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-158-383 Supplier ZeptoMetrix Supplier No. 0804442
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Infectious, titered microorganisms can be utilized for analytical testing during assay development, including limit of detection (LOD) and cross-reactivity studies. When used as a control for nucleic acid tests, the same protocols as those used to extract and amplify clinical specimens should be employed.

Each frozen aliquot contains 1 mL of a pure, titered culture of Proteus penneri. The identification of this organism was confirmed by 16S sequencing. The purity of the culture was monitored by Gram staining and by additional culturing. The titer was performed on a minimum of two aliquots after freezing. The freezing medium contains 15% glycerol as a cryoprotectant. Please see the Certificate of Analysis for the specific freezing medium used.

INTENDED USE:

Live, titered microorganisms can be used to determine a limit of detection (LOD), in diagnostic assay development or cross reactivity studies. When used as a control for nucleic acid tests, the same protocols as those used to amplify clinical specimens should be employed.

Specifications

Content And Storage 2°C to 8°C
Description Can be used to determine a limit of detection (LOD), in diagnostic assay development or cross reactivity studies
Quantity 1 mL
Regulatory Status RUO
Strain Designations Proteus penneri Z527
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.