Learn More
ATCC Proteus vulgaris
Catalog No. 502381955
Encompass
Due to an upcoming ERP system upgrade, ATCC will have a temporary system shutdown from 11/20/26-12/1/26. All orders will experience a 5-6 days delay in processing. Orders will continue to be sent to ATCC during the blackout and will be held until 12/2/26 when they are released. Any questions, please reach out to Encompass.Supplierchannel@thermofisher.com.
Proteus vulgaris is a whole-genome sequenced bacterium that has applications as a quality control strain for Autobac and Roche Diagnostics products and the VITEK 2 GN Comprehensive QC Set.
Catalog No. 50-238-1955 Supplier ATCC Supplier No. 6380
Due to an upcoming ERP system upgrade, ATCC will have a temporary system shutdown from 11/20/26-12/1/26. All orders will experience a 5-6 days delay in processing. Orders will continue to be sent to ATCC during the blackout and will be held until 12/2/26 when they are released. Any questions, please reach out to Encompass.Supplierchannel@thermofisher.com.
Proteus vulgaris is a whole-genome sequenced bacterium that has applications as a quality control strain for Autobac and Roche Diagnostics products and the VITEK 2 GN Comprehensive QC Set.