Learn More
ATCC Proteus vulgaris
Supplier:  ATCC 49132
Encompass
Proteus vulgaris strain AmMS 105 is a whole-genome sequenced clinical isolate that can be used as a quality control strain for MicroScan products.
Catalog No. 50-238-4246